SY3H3D67B06F Synthonix Corporation

2-(2-Chloro-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • Cas Number : 1256781-74-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57908.html

Safety Information

MSDS https://synthonix.com/57908.html
Availability details for
Quantity
  • Availability
  • Add to Cart