SY3H3D67B071 Synthonix Corporation

4-Chloro-2-phenylthieno[3,2-c]pyridine-7-carbonitrile

  • MDL number MFCD21605514
  • Cas Number : 1361197-82-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58023.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58023.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart