SY3H3D67B846 Synthonix Corporation

2-Chloro-N-(2-(2-chlorobenzoyl)-4-nitrophenyl)acetamide

  • MDL number MFCD00799975
  • Cas Number : 180854-85-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58111.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58111.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart