SY3H3D67B84A Synthonix Corporation

N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-nitroquinazolin-4-amine

  • MDL number MFCD27975589
  • Cas Number : 179552-74-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58119.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58119.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart