SY3H3D67B84C Synthonix Corporation

N-(2-Chlorophenyl)-1H-indole-3-carboxamide

  • MDL number MFCD12478724
  • Cas Number : 61788-27-0
  • Molecular Weight : 270.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58136.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/58136.html
pictograms GHS07
hstatements H302, H315, H319, H332, H335
Availability details for
Quantity
  • Availability
  • Add to Cart