SY3H3D67B84F Synthonix Corporation

3-(2-Chloropyrimidin-4-yl)-1-cyclopropyl-1H-indole

  • MDL number MFCD30723492
  • Cas Number : 1899922-76-9
  • Molecular Weight : 269.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Yellow solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58155.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58155.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart