SY3H3D67B856 Synthonix Corporation

2-Chloro-N-(3-(2-chlorobenzoyl)-5-ethylthiophen-2-yl)acetamide

  • MDL number MFCD18969310
  • Cas Number : 50508-89-9
  • Molecular Weight : 342.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Light-brown to brown powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58195.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58195.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart