SY3H3D67B859 Synthonix Corporation

1-Chloro-2-(4-ethoxybenzyl)-4-iodobenzene

  • MDL number MFCD18642361
  • Cas Number : 1103738-29-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58248.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58248.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart