SY3H3D67B86B Synthonix Corporation

4-Chloro-2,6-diphenylpyrimidine

  • MDL number MFCD00234890
  • Cas Number : 29509-91-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Pale-yellow or Gray
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58567.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58567.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart