SY3H3D67B86C Synthonix Corporation

(5-Chlorobenzofuran-2-yl)(4-methoxyphenyl)methanone

  • Cas Number : 220526-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58579.html

Safety Information

MSDS https://synthonix.com/58579.html
Availability details for
Quantity
  • Availability
  • Add to Cart