SY3H3D67B86E Synthonix Corporation

4-Cyano-3-fluorophenyl 4-ethylbenzoate

  • MDL number MFCD00689047
  • Cas Number : 86776-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58588.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58588.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart