SY3H3D67B879 Synthonix Corporation

4-((6-((3-Chloropropanoyl)oxy)hexyl)oxy)benzoic acid

  • Cas Number : 182922-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58769.html

Safety Information

MSDS https://synthonix.com/58769.html
Availability details for
Quantity
  • Availability
  • Add to Cart