SY3H3D67B87B Synthonix Corporation

7-Chloro-6-fluoro-1-(4-fluorophenyl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

  • Cas Number : 98105-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58872.html

Safety Information

MSDS https://synthonix.com/58872.html
Availability details for
Quantity
  • Availability
  • Add to Cart