SY3H3D67B884 Synthonix Corporation

(S)-(2-Chloro-5-iodophenyl)(4-((tetrahydrofuran-3-yl)oxy)phenyl)methanone

  • MDL number MFCD22665921
  • Cas Number : 915095-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58914.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58914.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart