SY3H3D67B892 Synthonix Corporation

3-Cyclopropyl-1-(2-fluoro-4-iodophenyl)-5-hydroxy-6,8-dimethylpyrido[2,3-d]pyrimidine-2,4,7(1H,3H,8H)-trione

  • Cas Number : 871700-24-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/59180.html

Safety Information

MSDS https://synthonix.com/59180.html
Availability details for
Quantity
  • Availability
  • Add to Cart