SY3H3D67B89C Synthonix Corporation

(S)-1-Cbz-3-Boc-Aminopiperidine

  • MDL number MFCD11501178
  • Cas Number : 876379-22-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59297.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59297.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart