SY3H3D67B89F Synthonix Corporation

N-(4-Chloro-2,5-dimethoxyphenyl)-3-hydroxy-2-naphthamide

  • MDL number MFCD00043903
  • Cas Number : 4273-92-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white to Pale-gray powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59371.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59371.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart