SY3H3D67B8A2 Synthonix Corporation

4-Cyano-3-fluorophenyl 4-pentylbenzoate

  • Cas Number : 86786-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/59396.html

Safety Information

MSDS https://synthonix.com/59396.html
Availability details for
Quantity
  • Availability
  • Add to Cart