SY3H3D67B8A3 Synthonix Corporation

6-Chloro-7,8-dimethoxy-1-(4-methoxyphenyl)-2,3,4,5-tetrahydro-1H-benzo[d]azepine

  • MDL number MFCD08460175
  • Cas Number : 67287-53-0
  • Molecular Weight : 347.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59420.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59420.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart