SY3H3D67B8A5 Synthonix Corporation

(E)-1-(3-(2-(7-Chloroquinolin-2-yl)vinyl)phenyl)prop-2-en-1-ol

  • MDL number MFCD09833496
  • Cas Number : 149968-10-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59496.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59496.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart