SY3H3D67B8A6 Synthonix Corporation

(Chloro(p-tolyl)methylene)dibenzene

  • MDL number MFCD01073758
  • Cas Number : 23429-44-9
  • Molecular Weight : 292.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to yellow or pale-red powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59501.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59501.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart