SY3H3D67B8AB Synthonix Corporation

1-Cyclopropyl-8-ethoxy-6-fluoro-7-(3-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

  • Cas Number : 182868-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/59565.html

Safety Information

MSDS https://synthonix.com/59565.html
Availability details for
Quantity
  • Availability
  • Add to Cart