SY3H3D67B8B7 Synthonix Corporation

4,6-Dihydroxy-2-methylpyrimidine

  • MDL number MFCD00006104
  • Cas Number : 40497-30-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to Yellow Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39176.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39176.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart