SY3H3D67B8EA Synthonix Corporation

1,4-Dimethyl-2-nitrobenzene

  • Cas Number : 89-58-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/39515.html

Safety Information

MSDS https://synthonix.com/39515.html
Availability details for
Quantity
  • Availability
  • Add to Cart