SY3H3D67B8F2 Synthonix Corporation

3-(2,6-Dichlorophenyl)acrylic acid

  • MDL number MFCD00004374
  • Cas Number : 5345-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to Very pale yellow Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39564.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39564.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart