SY3H3D67B8F5 Synthonix Corporation

2-(2,5-Dimethylphenyl)acetic acid

  • MDL number MFCD00014369
  • Cas Number : 13612-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Very pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39606.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39606.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart