SY3H3D67B907 Synthonix Corporation

2,2'-Dimethyl-[1,1'-biphenyl]-4,4'-diamine

  • MDL number MFCD02269175
  • Cas Number : 84-67-3
  • Molecular Weight : 212.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39725.html

Safety Information

signalword Warning
pcodes P202, P261, P280
MSDS https://synthonix.com/39725.html
pictograms GHS07
hstatements H315, H319
Availability details for
Quantity
  • Availability
  • Add to Cart