SY3H3D67B90A Synthonix Corporation

(3,4-Dimethoxyphenyl)(phenyl)methanone

  • MDL number MFCD00075875
  • Cas Number : 4038-14-6
  • Molecular Weight : 242.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39731.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39731.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart