SY3H3D67B90C Synthonix Corporation

4-(3,4-Dichlorophenyl)-3,4-dihydronaphthalen-1(2H)-one

  • MDL number MFCD02093088
  • Cas Number : 79560-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Very pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39734.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39734.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart