SY3H3D67B93D Synthonix Corporation

2,5-Diamino-4,6-dichloropyrimidine

  • MDL number MFCD00792652
  • Cas Number : 55583-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Very pale yellow to pale red to red to brown or gray powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40098.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40098.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart