SY3H3D67B941 Synthonix Corporation

4,6-Diamino-5-nitropyrimidine

  • MDL number MFCD00023268
  • Cas Number : 2164-84-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40155.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40155.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart