SY3H3D67B946 Synthonix Corporation

2,5-Diaminopyrimidine-4,6-diol

  • Cas Number : 40769-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40206.html

Safety Information

MSDS https://synthonix.com/40206.html
Availability details for
Quantity
  • Availability
  • Add to Cart