SY3H3D67B949 Synthonix Corporation

1,4-Dimethyl-1H-1,2,3-triazole

  • Cas Number : 60166-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/40221.html

Safety Information

MSDS https://synthonix.com/40221.html
Availability details for
Quantity
  • Availability
  • Add to Cart