SY3H3D67B960 Synthonix Corporation

2,2-Dimethylmalonamide

  • Cas Number : 41882-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/40372.html

Safety Information

MSDS https://synthonix.com/40372.html
Availability details for
Quantity
  • Availability
  • Add to Cart