SY3H3D67B98B Synthonix Corporation

3,5-Dibromo-2-chloropyridin-4-amine

  • MDL number MFCD11110699
  • Cas Number : 1054484-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40560.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40560.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart