SY3H3D67B992 Synthonix Corporation

2,4-Dichloro-6-methyl-5-nitropyrimidine

  • MDL number MFCD00023196
  • Cas Number : 13162-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Pale-yellow to brown solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40610.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40610.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart