SY3H3D67B9A5 Synthonix Corporation

2,6-Difluoropyridin-3-ol

  • MDL number MFCD11044351
  • Cas Number : 209328-85-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40683.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40683.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart