SY3H3D67B9AE Synthonix Corporation

3,5-Dichloropyridin-2-amine

  • MDL number MFCD00006313
  • Cas Number : 4214-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40722.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40722.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart