SY3H3D67B9B2 Synthonix Corporation

4,5-Dichloro-2-methylpyrimidine

  • Cas Number : 26740-71-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/40727.html

Safety Information

MSDS https://synthonix.com/40727.html
Availability details for
Quantity
  • Availability
  • Add to Cart