SY3H3D67B9B3 Synthonix Corporation

4,5-Dichloro-6-methylpyrimidine

  • MDL number MFCD09909763
  • Cas Number : 83942-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40728.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40728.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart