SY3H3D67B9BD Synthonix Corporation

3,5-Dibromo-1-methylpyrazin-2(1H)-one

  • MDL number MFCD12022617
  • Cas Number : 87486-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Off white to Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40769.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40769.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart