SY3H3D67B9C1 Synthonix Corporation

4,5-Dichloro-1H-pyrrole-2-carboxamide

  • MDL number MFCD20542609
  • Cas Number : 50371-68-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40774.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40774.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart