SY3H3D67B9CD Synthonix Corporation

2,4-Dihydroxy-5-methoxypyrimidine

  • MDL number MFCD00187903
  • Cas Number : 6623-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Off white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41043.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41043.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart