SY3H3D67B9D9 Synthonix Corporation

5,6-Diamino-1-methyluracil

  • Cas Number : 6972-82-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41212.html

Safety Information

MSDS https://synthonix.com/41212.html
Availability details for
Quantity
  • Availability
  • Add to Cart