SY3H3D67B9E4 Synthonix Corporation

4,4-Difluoropiperidine

  • MDL number MFCD03094281
  • Cas Number : 21987-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41254.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41254.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart