SY3H3D67B9EF Synthonix Corporation

4,6-Dichloropyrimidine-2-carbonitrile

  • Cas Number : 1451391-83-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41291.html

Safety Information

MSDS https://synthonix.com/41291.html
Availability details for
Quantity
  • Availability
  • Add to Cart