SY3H3D67B9F1 Synthonix Corporation

3,5-Dichloro-2,6-difluoropyridine

  • MDL number MFCD03001157
  • Cas Number : 698-51-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41295.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41295.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart