SY3H3D67B9FB Synthonix Corporation

N,1-Dimethyl-1H-pyrazol-4-amine dihydrochloride

  • MDL number MFCD19689724
  • Cas Number : 948572-94-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41402.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41402.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart