SY3H3D67B9FD Synthonix Corporation

1,4-Diformylpiperazine

  • MDL number MFCD00006153
  • Cas Number : 4164-39-0
  • Molecular Weight : 142.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form powder Colour: light yellow - yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41433.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41433.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart