SY3H3D67BA04 Synthonix Corporation

5,6-Diamino-3-methyl-2-(methylthio)pyrimidin-4(3H)-one

  • MDL number MFCD02091208
  • Cas Number : 39008-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41451.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41451.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart